Order Entry
Austria
ContactUsLinkComponent
Tork Skincare Dispenser - with Intuition™ Sensor
Tork Skincare Dispenser - with Intuition™ Sensor
Catalog # SCAA561608
Supplier:  Essity
New Product
undefined
Tork Skincare Dispenser - with Intuition™ Sensor
Catalog # SCAA561608
Supplier:  Essity
New Product

Specifications

  • Description:
    Tork skincare dispenser - with Intuition™ sensor, plastic, S4 - foam soap system, black
  • Pk:
    1

Specifications

About this item

The Tork skincare dispenser with Intuition™ sensor in elevation design not only accommodates a wide range of Tork skincare products in sealed bottles but also leaves a lasting impression on your guests.

  • Sensor driven, touch-free dispensing reduces the risk of cross contamination
  • For all: Proven easy-to-use design by third-party certificate* – promotes good hand hygiene for all users, even children and the elderly
  • Reduce time for cleaning staff: Certified effortless refilling * in less than 10 s
  • High-capacity reducing waste: Helps reduce consumption by up to 50%
  • W×D×H: 112×128×278 mm

Helps secure good hygiene: Factory-sealed bottle with a new pump for every refill helps reduce risk of contamination and protect the formulations all the way to the user.

Its touch-free dispensing ensures good hand hygiene for all users. Elevation dispensers feature a functional and modern design that enhances any environment.