Order Entry
Austria
ContactUsLinkComponent
Lead Standard Solution, 1000 mg/L in Diluted HNO₃ for ICP/MS
Lead Standard Solution, 1000 mg/L in Diluted HNO₃ for ICP/MS
Catalog # ACSDM241.2NP.L1
Supplier:  CPACHEM
New Product
CAS Number:  
Lead Standard Solution, 1000 mg/L in Diluted HNO₃ for ICP/MS
Catalog # ACSDM241.2NP.L1
Supplier:  CPACHEM
New Product
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Pk:
    100 mL
  • Description:
    Lead [Pb] 1000 mg/L [HNO₃] Nitric acid 2 %

Specifications

About this item

Certifications: ISO 17034, 17025, 9001 certified.