Order Entry
Austria
ContactUsLinkComponent
Holmium Oxide Solution for Wavelength Control
Holmium Oxide Solution for Wavelength Control
Catalog # ACSDUV2225WL.L03
Supplier:  CPACHEM
New Product
CAS Number:  
Holmium Oxide Solution for Wavelength Control
Catalog # ACSDUV2225WL.L03
Supplier:  CPACHEM
New Product
CAS Number:  

Specifications

  • Pk:
    30 mL
  • Description:
    Holmium oxide solution for wavelength control acc. 2.2.25 of Ph Eur
  • Storage conditions:
    Store under normal laboratory conditions
  • Shelf life:
    36 Months
  • Storage temperature:
    15...25 °C

Specifications

About this item

Certifications: ISO 9001 certified.