Order Entry
Austria
Orders LinkContactUsLinkComponent
Ammonia solution, Test Solution (TS)

Ammonia solution, Test Solution (TS)

1 L
Catalog # ACSDCH8002013.1L
Supplier:   CPACHEM
Ammonia solution, Test Solution (TS)
Catalog # ACSDCH8002013.1L
Supplier:  CPACHEM
CAS Number:  1336-21-6
Ammonia solution, Test Solution (TS)

1 L

CAS Number:  1336-21-6
Price
91,00 €
Log in for availability
1 * 1 L
Product Details & Documents

Certifications: ISO 9001 certified.

Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Storage Temperature:Room Temperature
Specifications

Specifications

  • Pk:
    1 L

Specifications