Order Entry
Austria
ContactUsLinkComponent
Cadmium Standard Solution, 1 g/L Cd in dil. Nitric Acid for AAS
Cadmium Standard Solution, 1 g/L Cd in dil. Nitric Acid for AAS
Catalog # ACSDA010.2NP.L1
Supplier:  CPACHEM
New Product
CAS Number:  
Cadmium Standard Solution, 1 g/L Cd in dil. Nitric Acid for AAS
Catalog # ACSDA010.2NP.L1
Supplier:  CPACHEM
New Product
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Description:
    Cadmium standard solution, 1 g/L Cd in dil. nitric acid for AAS
  • Storage conditions:
    Store under normal laboratory conditions
  • Hazard class description:
    Corrosive substances
  • UN No.:
    1760
  • GHS signal:
    Danger
  • Storage temperature:
    15...25 °C
  • Physical form:
    Fluid
  • Element:
    Cadmium
  • Symbol:
    Cd

Specifications

About this item

Ordering information: Shelf life on ship date: 36 months - Store under normal laboratory conditions, at temperatures between 15 to 25 °C.