Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Food Standards
Capstone Product A
Capstone Product A
Capstone Product A
EHERC11041290
:
EHRENSTORFER
New Product
:
Capstone Product A
EHERC11041290
:
EHRENSTORFER
New Product
:
Pk:
10 mg
CAS No.:
80475-32-7
Description:
Capstone product A, molecular weight: 528,33
Storage conditions:
20 °C