Order Entry
Austria
ContactUsLinkComponent
Single-Port Portable Parallel Analyser (PPA), SL250
Single-Port Portable Parallel Analyser (PPA), SL250
Catalog # HACHLPV4439911002
Supplier:  Hach
New Product
Single-Port Portable Parallel Analyser (PPA), SL250
Catalog # HACHLPV4439911002
Supplier:  Hach
New Product

Specifications

  • Model:
    SL250
  • Detector:
    Silicon photodiode
  • Protection class:
    III
  • Temperature range:
    5...50 °C
  • Description:
    Single-port portable parallel analyser (PPA)
  • Enclosure rating:
    IP 54
  • Electrical:
    100 - 240 VAC, 50/60 Hz
  • Battery life:
    >200 Chemkey tests per full battery charge
  • Data logging capacity:
    Memory: 1000 measured values
  • Interfaces:
    Mini USB port
  • Power supply:
    2,5 mm jack
  • Weight:
    1 kg
  • Pk:
    1

Specifications

About this item

One portable instrument - all the key parameters.

  • Probe-based tests include pH, dissolved oxygen, conductivity, and more
  • 5-Pin input connector: Two M12 connectors for IntelliCal™ probes
  • 130,8×58,9×258,3 mm dimension

Hach's SL250 Portable parallel analyser (PPA) is an innovative multiparameter water quality meter that allows you to simultaneously perform colorimetric and probe-based measurements. The instrument and corresponding Chemkey reagents are easy to use regardless of technical expertise. Compared to traditional powder or liquid reagent methods, Chemkeys use a simplified testing method that requires no sample preparation, making the process significantly easier, faster, and safer while drastically reducing variability in results.

Colorimetric tests include free chlorine, total chlorine, fluoride, high range manganese, zinc, copper, nitrite, dissolved iron, high and low range alkalinity, high and low range orthophosphate, high and low range hardness, peracetic acid, and more.

Certifications: UKCA, FCC, ISED, RCM, KC.
Delivery: SL250 PPA with carry case, 125 V power cable and manual resources.