ContactUsLinkComponent
BasicKjel Distillation Unit
BasicKjel Distillation Unit
Catalog # BUCH11K36521110
Supplier:  BUCHI
New Product
BasicKjel Distillation Unit
Catalog # BUCH11K36521110
Supplier:  BUCHI
New Product
Specifications
  • Description:
    BasicKjel distillation unit with glass protection vessel and without chiller
  • Pk:
    1
Specifications
About this item
For protein and nitrogen determination (TKN, NPN, TVBN, ammonia, nitrate/nitrite and casein). Possibility to connect Metrohm or SI analytics titrators for the MultiKjel.
Three possible equipment:
EasyKjel: Basic unit
BasicKjel: Intermediate unit
MultiKjel: Advanced unit
Two types of protection: Plastic or glass protective vase: To protect the condenser from splashes caused by the distillation of water vapour.