Order Entry
Austria
ContactUsLinkComponent
 
DNA 20 kb Plasmid and Linear Kit
  EKSI5311708
 :  EKSIGENT
New Product
 :  
DNA 20 kb Plasmid and Linear Kit
  EKSI5311708
 :  EKSIGENT
New Product
 :  

 

  • Pk:
    1 KIT
  • Application:
  • Method / Format:
  • Refine target molecule:
  • Sample type:
  • Target molecule:
  • Description:
    DNA 20 kb plasmid and linear kit
  • Environmentally Preferable:

 

 

The DNA 20 kb plasmid and linear kit is used to do isoform and purity analysis of plasmid and linearised dsDNA.

This kit supplies high analytical resolution and can analyse a wide range of linear double-stranded DNA fragments or plasmid isoforms. Plasmid isoforms from 2,000 bp to 19,000 bp can be separated, including super coiled, linear, and open circular species. The kit can do size estimation and purity analysis for linear dsDNA samples from 100 bp to 20,000 bp.

 : This kit provides reagent volumes for analysis of 200 samples and includes: Acid wash/regenerating solution (0,1 N HCl) (125 ml)- 1, CE grade water (140 ml)- 2, DNA 20 kb plasmid and linear conditioning solution (135 ml)- 1, DNA 20 kb plasmid and linear gel (135 ml)- 1, DNA 20 kb plasmid and linear sample buffer (55 ml)- 1, SYBR™ gold nucleic acid gel stain1 (120 µl)- 6, DNA 20 kb plasmid test mix (8 ng in 20 µl)- 6 1 SYBR™ is a trademark of the life technologies corporation. SYBR™ Gold nucleic acid gel stain is not available for resale.