Order Entry
Austria
ContactUsLinkComponent
BioTracker Far-red Labile Fe2+ Dye
BioTracker Far-red Labile Fe2+ Dye
Catalog # SIALSCT037-1KIT
Supplier:  Merck
New Product
CAS Number:  
BioTracker Far-red Labile Fe2+ Dye
Catalog # SIALSCT037-1KIT
Supplier:  Merck
New Product
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    Fe²⁺ dye
  • Detection method:
    Fluorometric
  • Application:
    Cell analysis
  • Colour:
    White to blue
  • Format:
    Lyophilised (solid blue)

Specifications

About this item

Live cell imaging probe for labile ferrous (Fe²⁺) ions, localising inside endoplasmic reticulum (ER). Also suitable for flow cytometry.

BioTracker far-red labile Fe²⁺ dye is a fluorescent probe that specifically detects labile iron (II) ions (Fe²⁺). This probe is designed to selectively react with only Fe²⁺ separately from other metal ions and irreversibly turns into a far-red fluorescent substance. It mainly localises inside endoplasmic reticulum (ER) and/or golgi bodies. It can also be used with a flow cytometer equipped with a red laser. FerroFarRed shows almost no cytotoxicity at concentrations up to 100 μM (~20 times that of generally used concentrations).

Caution: Research use only and are not to be used for any other purpose.