VWR®, Arsenic standard solution, 1,000 mg/L As in dil. nitric acid AVS TITRINORM standard for AAS
- Traceable to SRM from NIST
- Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
- Supplied with certificate of analysis, real value ±0,3%
- Environmentally Preferable:
Frequently Bought Together