Wright's stain for microscopical staining
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Synonyms: Eosin Methylene blue solution according to Wright, Wright's eosin methylene blue
- CAS Number:68988-92-1
- MDL Number:MFCD00082143
- Density:0.8 g/cm³ (20 °C)
2 Options Available Below
Product Details & Documents
Synonyms: Eosin Methylene blue solution according to Wright, Wright's eosin methylene blue
- CAS Number:68988-92-1
- MDL Number:MFCD00082143
- Density:0.8 g/cm³ (20 °C)



