Order Entry
Austria
ContactUsLinkComponent
4-Nitrophenyl phosphate disodium salt hexahydrate (pNPP)
4-Nitrophenyl phosphate disodium salt hexahydrate (pNPP)
Catalog # MOLE35359763-25G
Supplier:  Molekula
CAS Number:  
4-Nitrophenyl phosphate disodium salt hexahydrate (pNPP)
Catalog # MOLE35359763-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
Melting Pt:300 °C
 :371.144 g/mol
Storage Temperature:Freezer
EINECS:224-246-5
CAS Number:333338-18-4
 :4264-83-9
Formula:C₆H₄NNa₂O₆P·6H₂O
MDL Number:MFCD00066288