Order Entry
Austria
ContactUsLinkComponent
4-Nitrophenyl phosphate disodium salt hexahydrate (pNPP), tablets
4-Nitrophenyl phosphate disodium salt hexahydrate (pNPP), tablets
Catalog # CAYM10009147-1
Supplier:  Cayman Chemical
CAS Number:  
4-Nitrophenyl phosphate disodium salt hexahydrate (pNPP), tablets
Catalog # CAYM10009147-1
Supplier:  Cayman Chemical
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
Melting Pt:300 °C
 :371.144 g/mol
Storage Temperature:Freezer
EINECS:224-246-5
CAS Number:333338-18-4
 :4264-83-9
Formula:C₆H₄NNa₂O₆P·6H₂O
MDL Number:MFCD00066288