
N-Octyl-β-D-glucopyranoside ≥98%, white powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:29836-26-8
:
Formula:C₁₄H₂₈O₆
MDL Number:MFCD00063288
:292.37 g/mol
Melting Pt:89…100 °C (lit.)
EINECS:249-887-8
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Identity Test:Passes
- Purity:≥98%
- FTIR:Conforms to Standard
- Optical Rotation (c=5, water):-31 ± 3°
Specification Test Results
Frequently Bought Together