
N-Octyl-β-D-glucopyranoside 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:29836-26-8
:
Formula:C₁₄H₂₈O₆
MDL Number:MFCD00063288
:292.37 g/mol
Melting Pt:105 °C
EINECS:249-887-8
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to off-white Powder and/or chunks
- Infrared spectrum:Conforms
- Solubility:(10 % in water) Clear to slightly hazy colorless to light yellow
- HPLC:≥97.5 % (ELSD)
- Specific optical rotation:-31° to -35° (20°C, 589 nm) (c=5, H2O)
Specification Test Results
Frequently Bought Together