Order Entry
Austria
ContactUsLinkComponent
Sodium chloride regeneration salt, BROXO for water softening
Sodium chloride regeneration salt, BROXO for water softening
Catalog # 148-2505
Supplier:  KAMMERER
CAS Number:  
Sodium chloride regeneration salt, BROXO for water softening
Catalog # 148-2505
Supplier:  KAMMERER
CAS Number:  

Specifications

  • Pk:
    25 kg
  • Seq:
    4283

Specifications

About this item

 :
 :58.44 g/mol
Boiling Pt:1416 °C (1013 hPa)
Melting Pt:801 °C
Density:2.17 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-598-3
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477