Order Entry
Austria
ContactUsLinkComponent
Formalin 10%, buffered (neutral) (pH 7.0 ± 0.2), Sigma-Aldrich®
Formalin 10%, buffered (neutral) (pH 7.0 ± 0.2), Sigma-Aldrich®
Catalog # HT501128-4L
CAS Number:  
Formalin 10%, buffered (neutral) (pH 7.0 ± 0.2), Sigma-Aldrich®
Catalog # HT501128-4L
CAS Number:  

Specifications

  • Pk:
    4 L
  • Seq:
    5162

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ200
 :30.03 g/mol
EINECS:200-001-8
 :GHS08
 :GHS07
Storage Temperature:Ambient
CAS Number:50-00-0
Formula:CH₂O
MDL Number:MFCD00003274