

Copper(II) sulphate pentahydrate, Sigma-Aldrich®
Catalog # SIALC3036-250G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g
- Seq:3545
Specifications
About this item
:
:249.69 g/mol
Merck Index:13,02682
Density:2.284 g/cm³ (20 °C)
ADR:9,III
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:231-847-6
UN:3077
REACH:01-2119520566-40
:GHS07
:GHS09
CAS Number:7758-99-8
Formula:CuSO₄·5H₂O
MDL Number:MFCD00149681