Dicyclohexylcarbodiimide
Glass bottle for solids, 1 kg
Catalog # MOLE57041553-1KG
Marketsource
Supplier: Molekula
Dicyclohexylcarbodiimide
Catalog # MOLE57041553-1KG
Supplier: Molekula
Marketsource
CAS Number: 538-75-0
Jump to:
Product Details & Documents
Synonyms: 1,3-Dicyclohexylcarbodiimide, DCC, N,N'-Dicyclohexylcarbodiimide
- CAS Number:538-75-0
- Formula:C₁₃H₂₂N₂
- MDL Number:MFCD00011659
- Molecular Weight:206.33 g/mol
- Merck Index:13,03123
- Boiling Pt:148…152 °C (15 hPa)
- Melting Pt:35…36 °C
- Density:0.95 g/cm³ (40 °C)
- Flash Pt:113 °C
- EINECS:208-704-1
- UN:2811
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



