Tetrazolium blue chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1871-22-3
:
Formula:C₄₀H₃₂Cl₂N₈O₂
MDL Number:MFCD00040933
Merck Index:12,09380
Melting Pt:265…270 °C
:M-Clarity™ quality level = MQ200
:727.65 g/mol
Storage Temperature:Ambient
EINECS:217-488-8
Frequently Bought Together