Order Entry
Austria
ContactUsLinkComponent
(3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxy-1-propanesulphonate)
(3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxy-1-propanesulphonate)
Catalog # DG053
Supplier:  G-Biosciences
CAS Number:  
(3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxy-1-propanesulphonate)
Catalog # DG053
Supplier:  G-Biosciences
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :630.89 g/mol
Melting Pt:184…186 °C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:82473-24-3
Formula:C₃₂H₅₈N₂O₈S