(3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxy-1-propanesulphonate)
Catalog # DG052
Supplier: G-Biosciences
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:630.89 g/mol
Melting Pt:184…186 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:82473-24-3
Formula:C₃₂H₅₈N₂O₈S