ContactUsLinkComponent
Charcoal Agar Base Dehydrated Culture Media
Charcoal Agar Base Dehydrated Culture Media
Catalog # HIMEM344-500G
Supplier:  HIMEDIA
New Product
CAS Number:  
Charcoal Agar Base Dehydrated Culture Media
Catalog # HIMEM344-500G
Supplier:  HIMEDIA
New Product
CAS Number:  
Specifications
  • Pk:
    1
  • Description:
    Charcoal agar base, 500 g
Specifications
About this item
For the cultivation of Bordetella pertussis, vaccine production, and stock culture maintenance.
  • Microbiological culture media
  • 500 g poly bottles