Order Entry
Austria
ContactUsLinkComponent
Copper (II) ethylenediamine 1 mol/l (3 N) in aqueous solution for measuring the viscosity of cellulose acc. to DIN 54270, Supelco®
Corrosive
Irritant
Copper (II) ethylenediamine 1 mol/l (3 N) in aqueous solution for measuring the viscosity of cellulose acc. to DIN 54270, Supelco®
Catalog # 1.09288.1000
Supplier:  Merck
CAS Number:  
Corrosive
Irritant
Copper (II) ethylenediamine 1 mol/l (3 N) in aqueous solution for measuring the viscosity of cellulose acc. to DIN 54270, Supelco®
Catalog # 1.09288.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Density:
    1.10 g/cm3 (20 °C)
  • pH:
    12.0 - 12.5 (10 g/l, H2O, 20 °C)
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive material

Specification Test Results

About this item

 :
 :217.76 g/mol
Density:1.1 g/cm³ (20 °C)
ADR:8 (6.1),II
 :M-Clarity™ quality level = MQ100
UN:1761
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:14552-35-3
Formula:Cu(H₂NCH₂CH₂NH₂)₂(OH)₂
MDL Number:MFCD00274628