
EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride)
Catalog # 59002.
Supplier: Biotium
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:191.7 g/mol
Melting Pt:112…113 °C
EINECS:247-361-2
:GHS07
Storage Temperature:Freezer
CAS Number:25952-53-8
Formula:C₈H₁₇N₃·HCl
MDL Number:MFCD00012503