
EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:25952-53-8
:
Formula:C₈H₁₇N₃·HCl
MDL Number:MFCD00012503
:191.7 g/mol
Melting Pt:112…113 °C
:M-Clarity™ quality level = MQ300
EINECS:247-361-2
:GHS07
Storage Temperature:Freezer
Frequently Bought Together