Order Entry
Austria
ContactUsLinkComponent
EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride) 98+%
Toxic
Health
Environment
EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride) 98+%
Certificates
undefined
Toxic
Health
Environment
EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride) 98+%
Certificates

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    111 to 116 °C
  • Titration Argentometric:
    ≥98.0 %

Specification Test Results

Related Information

Frequently Bought Together