Order Entry
Austria
ContactUsLinkComponent
Potassium pyroantimonate, Test Solution (TS)
Potassium pyroantimonate, Test Solution (TS)
Catalog # ACSDCH8002106.L1
Supplier:  CPACHEM
New Product
CAS Number:  
Potassium pyroantimonate, Test Solution (TS)
Catalog # ACSDCH8002106.L1
Supplier:  CPACHEM
New Product
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :262.9 g/mol
CAS Number:12208-13-8
 :MFCD00150439
Formula:KSbO₃·3H₂O
MDL Number:MFCD00011415