Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Buffer Capsules

 :   Greisinger
Buffer Capsules
 
Buffer Capsules

 

 

Cost-effective alternative: Buffer capsules and working sets. With buffer capsules, you can produce a 100 ml buffer solution in the most cost-effective manner. A particular advantage of the capsules is that their storage life is several years longer than that of ready-to-use solutions.

The capsules are offered in a practical working set: The most cost-effective entry with everything you need for a successful start in pH measurement, including cleaning solution and KCL100 storage/replacement electrolyte.

Product Family Options

 

  • Description
    pH value
    Colour
    Pk
     
     
  • pH buffer capsules, GPH 4, amount: 5 capsules
    4,0
    Red
    1 SET
       
       
    1 * 1 SET
    1 * 1 SET

  • pH buffer capsules, GPH 7, amount: 5 capsules
    7,0
    Green
    1 SET
       
       
    1 * 1 SET
    1 * 1 SET