Unsupported neoprene gloves that provide broad chemical protection with enhanced comfort and tactility.
- MICROCHEM™ chemical barrier technology: Unique multi-layer rubber polymer and plastic laminate technology for extremely broad chemical protection
- Multi-hazard protection: AlphaTec 53-002 provides next generational chemical protection whilst delivering secondary hazard protection, like mechanical hazards
- Comfort and performance: Unique donning and doffing layer to allow ease of use
Extreme Chemical protection: AlphaTec® 53-002 gloves achieving 17 out of 18 chemicals on EN 374:2016 with 16 being at class 6>480 minutes and 16 out of 18> class 2 against ASTM F739 with a 10x lower limit to pass.
Elevated comfort and dexterity: Unique manufacturing process for optimised fit with raised diamond grip for secure performance in a wide range of scenarios.
Improved durability: They also possess EN 388-compliant abrasion resistance for better mechanical protection.
Specialized protection: And anti-static according to EN 1149-3 standards preventing workplace injuries.
Silicone free and raised diamond grip design.
Primary industries: Automotive, chemical, life sciences, recycling and waste disposal, utilities.
Recommended for:
Assembly and inspection of components
Oil, fluids and filter change
Painting or cleaning tools or robots
Chemical handling
Transferring liquids and solids
Chemical leaks
Spill or leakage cleanup
- Catalog No:
- 112-6655
- 112-6656
- 112-6654
- 112-6653
- 112-6652
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Material:
- Neoprene
- Neoprene
- Neoprene
- Neoprene
- Neoprene
- Finish:
- Raised Diamond
- Raised Diamond
- Raised Diamond
- Raised Diamond
- Raised Diamond
- Cuff style:
- Gauntlet
- Gauntlet
- Gauntlet
- Gauntlet
- Gauntlet
- Palm thickness:
- 0,44 mm
- 0,44 mm
- 0,44 mm
- 0,44 mm
- 0,44 mm
- EN norm:
- EN ISO 374-1:2016 Type A (ABCEFGHIJKLMNOPST), EN ISO 374-5 VIRUS, EN 421, EN 388:2016 (1111X)
- EN ISO 374-1:2016 Type A (ABCEFGHIJKLMNOPST), EN ISO 374-5 VIRUS, EN 421, EN 388:2016 (1111X)
- EN ISO 374-1:2016 Type A (ABCEFGHIJKLMNOPST), EN ISO 374-5 VIRUS, EN 421, EN 388:2016 (1111X)
- EN ISO 374-1:2016 Type A (ABCEFGHIJKLMNOPST), EN ISO 374-5 VIRUS, EN 421, EN 388:2016 (1111X)
- EN ISO 374-1:2016 Type A (ABCEFGHIJKLMNOPST), EN ISO 374-5 VIRUS, EN 421, EN 388:2016 (1111X)
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Anti-static:
- +
- +
- +
- +
- +