Orders LinkContactUsLinkComponent
Extraction thimbles
Extraction thimbles
Supplier:   MACHEREY-NAGEL
Extraction thimbles
Supplier:  MACHEREY-NAGEL
Certificates
Extraction thimbles
Extraction thimbles
46 Options Available Below
Product Details & Documents
Extraction thimbles, made of pure cellulose, standard quality. With or without lid available.
  • Fat determination in food and feed
  • Determination of binder content in asphalt
  • Analysis of soil contaminants
  • Determination of additives in rubber and elastomers
Product Family Options
Grade-MN 645, without lid
Product Information
  • I-Ø×H
    Stärke
    Availability
    Price
  • 8×40 mm
    1,0 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 97,70 €
    97,70 €
    1 * 25 ST
       97,70 €
  • 9×50 mm
    1,0 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 97,70 €
    97,70 €
    1 * 25 ST
       97,70 €
  • 15×50 mm
    1,0 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 97,70 €
    97,70 €
    1 * 25 ST
       97,70 €
  • 15×100 mm
    1,0 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 97,70 €
    97,70 €
    1 * 25 ST
       97,70 €
  • 20×80 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 97,70 €
    97,70 €
    1 * 25 ST
       97,70 €
  • 22×80 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 97,70 €
    97,70 €
    1 * 25 ST
       97,70 €
  • 22×90 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 97,70 €
    97,70 €
    1 * 25 ST
       97,70 €
  • 23×100 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 97,70 €
    97,70 €
    1 * 25 ST
       97,70 €
  • 27× 60 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 97,70 €
    97,70 €
    1 * 25 ST
       97,70 €
  • 27× 80 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 97,70 €
    97,70 €
    1 * 25 ST
       97,70 €
  • 27×100 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 97,70 €
    97,70 €
    1 * 25 ST
       97,70 €
  • 28× 80 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 97,70 €
    97,70 €
    1 * 25 ST
       97,70 €
  • 28× 90 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 97,70 €
    97,70 €
    1 * 25 ST
       97,70 €
  • 28×100 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 97,70 €
    97,70 €
    1 * 25 ST
       97,70 €
  • 28×120 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 117,00 €
    117,00 €
    1 * 25 ST
       117,00 €
  • 29×100 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 117,00 €
    117,00 €
    1 * 25 ST
       117,00 €
  • 30× 60 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 97,70 €
    97,70 €
    1 * 25 ST
       97,70 €
  • 30× 80 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 97,70 €
    97,70 €
    1 * 25 ST
       97,70 €
  • 30× 90 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 119,00 €
    119,00 €
    1 * 25 ST
       119,00 €
  • 30×100 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 119,00 €
    119,00 €
    1 * 25 ST
       119,00 €
  • 30×150 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 148,00 €
    148,00 €
    1 * 25 ST
       148,00 €
  • 31×118 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 146,00 €
    146,00 €
    1 * 25 ST
       146,00 €
  • 31×130 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 146,00 €
    146,00 €
    1 * 25 ST
       146,00 €
  • 33× 80 mm
    2,0 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 107,00 €
    107,00 €
    1 * 25 ST
       107,00 €
  • 33× 94 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 119,00 €
    119,00 €
    1 * 25 ST
       119,00 €
  • 33×205 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 195,00 €
    195,00 €
    1 * 25 ST
       195,00 €
  • 34×120 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 146,00 €
    146,00 €
    1 * 25 ST
       146,00 €
  • 34×150 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 175,00 €
    175,00 €
    1 * 25 ST
       175,00 €
  • 38×200 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 228,00 €
    228,00 €
    1 * 25 ST
       228,00 €
  • 40×123 mm
    2,0 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 186,00 €
    186,00 €
    1 * 25 ST
       186,00 €
  • 40×150 mm
    2,0 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 186,00 €
    186,00 €
    1 * 25 ST
       186,00 €
  • 43×130 mm
    2,0 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 186,00 €
    186,00 €
    1 * 25 ST
       186,00 €
  • 48×145 mm
    2,0 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 223,00 €
    223,00 €
    1 * 25 ST
       223,00 €
  • 48×200 mm
    2,0 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 257,00 €
    257,00 €
    1 * 25 ST
       257,00 €
  • 48×230 mm
    2,0 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 261,00 €
    261,00 €
    1 * 25 ST
       261,00 €
  • 51×145 mm
    2,0 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 244,00 €
    244,00 €
    1 * 25 ST
       244,00 €
  • 51×180 mm
    2,0 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 266,00 €
    266,00 €
    1 * 25 ST
       266,00 €
  • 55×275 mm
    2,0 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 325,00 €
    325,00 €
    1 * 25 ST
       325,00 €
  • 57×315 mm
    2,0 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 385,00 €
    385,00 €
    1 * 25 ST
       385,00 €
  • 60×180 mm
    2,0 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 261,00 €
    261,00 €
    1 * 25 ST
       261,00 €
  • 68×250 mm
    2,0 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 346,00 €
    346,00 €
    1 * 25 ST
       346,00 €
  • 70×330 mm
    2,0 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 424,00 €
    424,00 €
    1 * 25 ST
       424,00 €
  • 75×160 mm
    2,0 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 294,00 €
    294,00 €
    1 * 25 ST
       294,00 €
Grade-MN 645 D, with lid
Product Information
  • I-Ø×H
    Stärke
    Availability
    Price
  • 30×80 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 173,00 €
    173,00 €
    1 * 25 ST
       173,00 €
  • 30×100 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 197,00 €
    197,00 €
    1 * 25 ST
       197,00 €
  • 31×130 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 235,00 €
    235,00 €
    1 * 25 ST
       235,00 €