Order Entry
Austria
ContactUsLinkComponent
cFMS Receptor Inhibitor IV ≥95% (by HPLC), Calbiochem®, Sigma-Aldrich®
cFMS Receptor Inhibitor IV ≥95% (by HPLC), Calbiochem®, Sigma-Aldrich®
  344039-5
 :  
cFMS Receptor Inhibitor IV ≥95% (by HPLC), Calbiochem®, Sigma-Aldrich®
  344039-5
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 mg
  • Seq:
    2210

 

 

 :
 :378.47 g/mol
 :M-Clarity™ quality level = MQ100
 :Freezer
 :959626-45-0
 :C₂₂H₂₆N₄O₂
 :MFCD10568148