SD-169 ≥97% (by HPLC), Calbiochem®, Sigma-Aldrich®
Catalog # 506158-10
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pack type:Plastic ampoule
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
:160.17 g/mol
:M-Clarity™ quality level = MQ100
Storage Temperature:Refrigerator
CAS Number:1670-87-7
Formula:C₉H₈N₂O
MDL Number:MFCD07779486