ContactUsLinkComponent
Precellys Multi-Tissue Dissociation Kit
Precellys Multi-Tissue Dissociation Kit
Catalog # BERTD05717.20EA
Supplier:  Bertin Instruments
New Product
CAS Number:  
Precellys Multi-Tissue Dissociation Kit
Catalog # BERTD05717.20EA
Supplier:  Bertin Instruments
New Product
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    Multi-tissue dissociation kit
Specifications
About this item
Optimise tissue dissociation: The Precellys Multi-Tissue Dissociation Kit developed by Bertin Technologies is an innovative and patented solution designed for the obtention of viable single cells from biological samples for research purposes. It enables the obtention of high-quality single cells from complex tissues such as mouse spleen, liver, lung, heart, and thymus and opens the door to downstream single-cell analysis, such as flow cytometry or NGS.
  • Versatile kit, compatible with a variety of tissues
  • High-yield of living cells
  • Compatible with Precellys Evolution and Precellys Evolution Touch homogenisers
Simplified process: This kit is specially designed to work with Precellys Evolution and Precellys Evolution Touch homogenisers. Thanks to the "bead-shearing" technology, it guarantees a high-yield recovery of viable single cells. Development studies showed that the percentage of viable cells isolated was over 80% for the most common tissue types used in this field and residual debris was minimised, subsequently simplifying the downstream analysis and the identification of the cell population of interest.
Versatile alternative to conventional processes: The Multi-Tissue Dissociation Kit is versatile, adapting to a large variety of tissues. Adjust the protocol to suit the specific sample - no need for special equipment or many sample-specific kits. This flexibility greatly facilitates sample preparation and optimises your research.