Orders LinkContactUsLinkComponent
GloCell™ Fixable Viability Dye Red 710
Marketsource
GloCell™ Fixable Viability Dye Red 710
Marketsource
GloCell™ Fixable Viability Dye Red 710
GloCell™ Fixable Viability Dye Red 710
Amine-labelling fluorescent dye for live/dead staining of mammalian cells - 100 tests.
GloCell™ Fixable viability dye red 710 is a fluorescent cell viability dye for staining live/dead mammalian cells in applications such as flow cytometry. The dye irreversibly binds intracellular and cell surface amine groups and can be used prior to fixation, permeabilisation, or cryopreservation. Cells with compromised plasma membranes become permeable to the GloCell™ dye, resulting in greater fluorescence compared to live cells in a cell viability assay.
GloCell™ Fixable viability dye red 710 is excited by the red laser (633 nm) with an excitation maximum of 647 nm, and it has a fluorescence emission maximum of 710 nm that can be detected using the 710/50 band pass filter used to detect Alexa Fluor® 700.
CAS No: 67-68-5
 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Product Family Options
  • Description
    Pk Info
    Pk
  • GloCell™ Fixable viability dye red 710
    100 Tests
    1 * 100 Tests
    1 * 100 Tests - 184,00 €
    184,00 €
    1 * 100 Tests
       184,00 €
    Marketsource
    Marketsource
  • GloCell™ Fixable viability dye red 710
    500 tests
    500 Tests
    1 * 500 Tests
    1 * 500 Tests - 710,00 €
    710,00 €
    1 * 500 Tests
       710,00 €
    Marketsource
    Marketsource