Chemical-Resistant PP Labels B-7425J for BradyJet™ J7300 Inkjet Printer
Supplier: Brady
Certificates
About this item
Create full-colour identification and barcode labels B-7425J that outlast liquid nitrogen, autoclave conditions, and harsh chemicals. For use on smooth surfaces.
- Labels withstand extreme temperatures from −80 to +70 °C
- Permanent acrylic adhesive bonds tightly to smooth surfaces like glass, copper, and stainless steel
- Inkjet receptive polypropylene material
- Designed for indoor use
- Abrasion-resistant, low temperature resistance, solvent/chemical resistant
- Matt finish
Inkjet receptive label material is ideal for full-colour laboratory identification on vials, centrifuge tubes, and test tubes. Built-in LabelSense™ technology enables cartridge and printer communication for fast changeovers, automatic label setup, and seamless software interaction.
For use with the BradyJet™ J7300 Colour Label Printer. Ink cartridges are required to print on this material; use part numbers P-CMY-1 and P-K-1.
Certifications: RoHS compliant to 2011/65/EU and 2015/863.
Frequently Bought Together