Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Bottles
Jerricans
Accessories for Jerricans, HDPE, with screw cap
Accessories for Jerricans, HDPE, with screw cap
Accessories for Jerricans, HDPE, with screw cap
SEMA9284
:
SEMADENI
New Product
Accessories for Jerricans, HDPE, with screw cap
SEMA9284
:
SEMADENI
New Product
For:
DIN 42, DIN 61
Colour:
Black
Description:
Lid for stacking carboy 55 mm
Pk:
1