Order Entry
Austria
ContactUsLinkComponent
Brady® Alignment Roll for BradyJet™ J7300 Colour Label Printer
Brady® Alignment Roll for BradyJet™ J7300 Colour Label Printer
  BRDY176579
 :  Brady
undefined
Brady® Alignment Roll for BradyJet™ J7300 Colour Label Printer
  BRDY176579
 :  Brady

 

  • Colour:
    White
  • Description:
    Alignment roll
  • Length:
    12,24 m
  • Pk:
    1
  • Width:
    101,60 mm

 

 

Use after replacing the ink cartridge for printhead alignment sequences.

  • For use with the BradyJet™ J7300 colour label printer