Quinoline for synthesis
SIAL8024070250
: Merck
New Product
:
- Pk:250 mL
- Seq:2590
- Environmentally Preferable:
- Colour (visual):Yellowish to yellowish brown to brownish
- Assay (GC, area%):≥97.0% (a/a)
- Density (d 20 °C/4 °C):1.092 - 1.094
- Identity (IR):Passes test
:
:129.16 g/mol
:2656
:GHS06
:GHS08
:GHS09
:–15 °C
:Room Temperature
:91-22-5
:C₉H₇N
:MFCD00006736
:237...238 °C/1013 hPa
:1.09 g/cm³ (25 °C)
:107 °C (closed cup)
:6.1 III
:13,08160
:M-Clarity™ quality level = MQ200