Order Entry
Austria
ContactUsLinkComponent
Human Untagged FBXL17 Clone
Human Untagged FBXL17 Clone
Catalog # ORIGSC327180
Supplier:  OriGene
New Product
CAS Number:  
Human Untagged FBXL17 Clone
Catalog # ORIGSC327180
Supplier:  OriGene
New Product
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Description:
    FBXL17 (NM_001163315) Human untagged clone
  • Vector details:
    pCMV6-Entry (PS100001)

Specifications

About this item

FBXL17 (untagged)-Human F-box and leucine-rich repeat protein 17 (FBXL17). The ORF clone is ion-exchange column purified and shipped in a 2D barcoded Matrix tube containing 10 µg of transfection-ready, dried plasmid DNA (reconstitute with 100 µl of water).

  • Centrifuge at 5000×g for 5 min
  • Carefully open the tube and add 100 µl of sterile water to dissolve the DNA
  • Close the tube and incubate for 10 minutes at room temperature
  • Briefly vortex the tube and then do a quick spin (less than 5000×g) to concentrate the liquid at the bottom
  • Store the suspended plasmid at −20 °C. The DNA is stable for at least one year from date of shipping when stored at −20 °C

E. coli Selection: Kanamycin (25 µg/ml)
Synonyms: Fbl17; Fbx13; FBXO13
Mammalian cell selection: Neomycin
Tag: Tag free