Self-Laminating Polyester Cryo Labels B-461 for 75 mm Core Printers
: Brady
Matte, permanent self-laminating and wrap-around B-461 polyester labels for smooth surfaces. For use on conical tubes, cryo tubes and vials. Suitable for autoclaves, hot wwater baths (100 °C) and cryo or liquid nitrogen applications.
- Die-cut labels are supplied on a roll
- Contains some adhesive backed wire marking materials, and also labels that are pre-printed with headers or templates
- Ultra-thin construction prevents jamming in racks and centrifuges
- Chemical-resistant, low temperature-resistant, smear-resistant
With small core for BBP11, BBP12, THT printers.
Compliant with Directive 2011/65/EU and 2015/863
: Compliant with Directive 2011/65/EU and 2015/863
: Supplied with the ribbon seperate, not in the same enclosure as the tape/sleeves
Frequently Bought Together