Grounding cable, type 23, 1,5 m
Catalog # SCTV108294
Supplier: S.C.A.T.
Jump to:
Product Details & Documents
Grounding cable for ground connection with MC grounding plug, 90° angled with clamp.
Specifications
- Description:Grounding cable, type 23
- Length:1,5 m
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



