Orders LinkContactUsLinkComponent
Thread Adapter, GL 32
Thread Adapter, GL 32
Thread adapter, Borosilicate glass 3.3, Transparent
Catalog # NEOLE-2933
Supplier:   NEOLAB
Thread Adapter, GL 32
Catalog # NEOLE-2933
Supplier:  NEOLAB
Thread Adapter, GL 32
Thread adapter, Borosilicate glass 3.3, Transparent
Price
46,49 €
Log in for availability
1 * 1 ST
Product Details & Documents
These are required to attach threaded vials to ground joint apparatus such as rotary evaporators.
Ordering information: Size-dependent screw couplings are required for this purpose.
Specifications
  • Colour:
    Transparent
  • Description:
    Thread adapter
  • Material:
    Borosilicate glass 3.3