Mouse Hepatitis Virus (MHV) ELISA Kit (Quantitative)
VWRU76578-234
New Product
:
- Pk:1
- Assay Type:Sandwich
- Host:Mouse
- Target protein:Hepatitis virus
- Description:Mouse Hepatitis virus(MHV) ELISA kit (quantitative)
- Sample type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Shelf life:6 months (4 °C); 12 months (–20 °C)
- Detection range:Positive/Negative
- Storage temperature:2…8 °C
- Sample volume:50 - 100 µl
- Sensitivity:-
- Regulatory status:RUO
- Assay Principle: Qualitative
- Research Topic: Microbiology
- Abbreviation: MHV
- Detection Wavelength: 450 nm