VWR® Sterile Nitrile Cleanroom Class 10 Gloves
Supplier: VWR Collection
Certificates
About this item
Powder-free gloves designed to provide comfort and tactile sensitivity.
- Hand-specific, beaded cuff
- Silicone-free
- Low particle count
- Specialized manufacturing process
VWR® Sterile nitrile cleanroom class 10 gloves are entirely latex-free to prevent the risk of allergic reactions. These gloves are supplied sterile.
Certifications: EN ISO 21420:2020, EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016; CAT III; AQL 1.5
Packaging: Each pair is double packaged in a wallet within a pouch, 20 pouches per bag, 10 bags per case (200 pairs).
Specifications
- Catalog No:
- 112-6211
- 112-6216
- 112-6217
- 112-6212
- 112-6213
- 112-6214
- 112-6215
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Sterility:
- Sterile
- Sterile
- Sterile
- Sterile
- Sterile
- Sterile
- Sterile