Human Serum Albumin (HSA) Powder-Low Endotoxin, 5 g
Catalog # GEMB800-126P-005
Supplier: GEMINI BIO-PRODUCTS
New Product
Specifications
- Sterility:Non sterile
- Description:Human serum albumin powder-low endotoxin
- Shipping conditions:Ships at ambient
- Storage temperature:Store refrigerated at 2...8 °C
- Pack type:Poly bottle
- Pk:5 g
Specifications
About this item
Human albumin is prepared by cold-alcohol fractionation from pooled human plasma and lyophilised. MW ~66 kDa. Endotoxin ≤2 EU/mg.
Caution: For cell culture, further manufacturing and research use only. Not for direct therapeutic use.