Order Entry
Austria
ContactUsLinkComponent
Spore Strip-Dual Species, Mesa Labs
Spore Strip-Dual Species, Mesa Labs
Catalog # MESL77-01-5-5100T
Supplier:  MESA LABORATORIES
Spore Strip-Dual Species, Mesa Labs
Catalog # MESL77-01-5-5100T
Supplier:  MESA LABORATORIES

Specifications

  • Ø:
    38 mm
  • Width:
    6 mm
  • Description:
    Spore Strip-Dual Species
  • Length:
    38 mm
  • Pk:
    100

Specifications

About this item

Spore Strips consist of a paper spore strip inoculated with a sterilization-resistant spore population which are packaged in a glassine envelope. The glassine envelope serves as a microbial barrier which protects the spore strip from post sterilization contamination. Reduced incubation time available with appropriate conditions.

  • Organisms: Geobacillus stearothermophilus 79531 and Bacillus atrophaeus 93721

Spore Strip Dual Species is a biological indicator used in monitoring the efficacy of Steam sterilization, Chemiclave® Sterilization, Ethylene Oxide sterilization, or Dry Heat sterilization processes.