Reference Material, Trifloxystrobin Metabolite NOA413163
EHERC17842400
New Product
:
- Pk:10 mg
- CAS No.:1367578-46-8
- Description:Trifloxystrobin metabolite NOA413163
- Shipping conditions:20 °C
- Formula:C₁₉H₁₅F₃N₂O₆
- Molecular weight:424,33
- Storage temperature:20 °C