ContactUsLinkComponent
HTRF 96-Well, White Plate, Low Volume
HTRF 96-Well, White Plate, Low Volume
  PERC66PL96005
New Product
HTRF 96-Well, White Plate, Low Volume
  PERC66PL96005
New Product
  • No. of wells:
    96
  • Colour:
    White
  • Description:
    HTRF 96-well plate, low volume, polystyrene
  • Dimensions:
    85,4W×127,6D×12,0H mm
  • Working volume:
    20 µl
  • Pk:
    5
  • Well volume:
    40 µl
These plates have been specifically designed as an alternative to 384-well plate small sample series or simply to try HTRF in a convenient environment.
  • HTRF with ELISA plate ease of use and low volume wells for low sample consumption
  • These 96-well low volume plates can be used with all HTRF assays
96-well low volume plates can be used with all HTRF assays, example second messengers, phospho-proteins, cytokines and biomarkers. you can be used in a no-transfer protocol with suspension cells or with a transfer step adherent cells. see plates can be read on all HTRF certified readers, thus making or implementation in your experiments really easy.
Well shape: Round
Well-bottom shape: V-bottom
 : For research use only. Not for use in diagnostic procedures.